Merck
HybriScan®
Quantification and Identification of Microorganisms in Beverages, Food and Water
HybriScan® Rapid Microbial Test System
Our HybriScan® molecular screening system uses sandwich hybridization to provide fast and accurate detection, identification and quantification of spoilage and pathogenic microorganisms in beverages, water and food. It is ideal for the comprehensive and reliable routine control of raw materials and concentrates in all production steps up to the quality check of finished goods.
The HybriScan® system can reduce your time to result by 2-14 days, depending on the target organism, while keeping costs low. Results are visible within 2 hours after the enrichment without the need for expensive instruments—the test can be performed with standard laboratory equipment and minimal training.
20-minuteTalk: In Situ Hybridization
Our expert, Jvo Siegrist, explains the technology of in situ hybridization on the example of the HybriScan® test system. Watch the presentation to learn about the advantages, disadvantages, and costs of this rapid method compared to common methods.
HybriScan® system screening workflow with analysis time of approx. 2–2.5 hours
Figure 1. HybriScan® system screening workflow with analysis time of approx. 2–2.5 hours
Rapid Microbiological Method: Sandwich Hybridization
The method simply needs a centrifuge, a thermal mixer and an optional a microplate reader. The complete analysis can be achieved in 2 hours for the HybriScan®D kit (quantitative kits) and about 1 hour for HybriScan®I kit (identification kits). Qualitative results can be determined by eye or, for quantitative measurement, use with a microplate reader.
Sensitivity, Specificity and Flexibility of Sandwich Hybridization
Sandwich hybridization is very sensitive and can detect attomoles of the target rRNA molecules in yeast or bacteria.1-3 These types of cells contain a large number of rRNA-containing ribosomes; a single cell therefore contains several thousand copies of rRNA but only one DNA copy. Sandwich hybridization provides sensitivity in crude biological samples because it is not susceptible to matrix interference. By using specific probes, the HybriScan® system allows both genus or species-specific detection and it is therefore applicable to many analytical fields, including monitoring the microbial content of beer, wine, non-alcoholic beverages, drinking water, a wide variety of foods and wastewater.
  • Detects viable microorganisms only—minimizes false-positive results from dead bacteria
  • Not sensitive to sample matrix
  • Genus- and species-specific detection
  • Results in just 2 hours after enrichment
  • Broad application range: beer, wine, non-alcoholic beverages, drinking water, food and wastewater
Advantages of HybriScan® technology over other microbial detection techniques
Hybriscan Molecular Detection
Figure 2.Hybriscan Molecular Detection
HybriScan® Waste Water Bacterial Count Calculator
For our Wastewater Kits (04447 HybriScan® D Waste Water Microthrix parvicella, 78436 HybriScan® D Waste Water Total Bacterial Count), we provide an excel sheet for calculation of total bacterial count, Microthrix count and ratio of Microthrix : Total Bacterial Count.
To download the file please click on the links below:
Hybriscan Waste Water Calculator English
Excel Sheet 28KB
Waste Water Calculator English
Hybriscan Waste Water Calculator Deutsch
Excel Sheet 56KB
Waste Water Calculator Deutsch
Materials
Loading
References
1.
Huhtamella S, Leinonen M, Nieminen T, Fahnert B, Myllykoski L, Breitenstein A, Neubauer P. 2007. RNA-based sandwich hybridisation method for detection of lactic acid bacteria in brewery samples. Journal of Microbiological Methods. 68(3):543-553. https://doi.org/10.1016/j.mimet.2006.10.009
2.
Leskelä T, Tilsala-Timisjärvi A, Kusnetsov J, Neubauer P, Breitenstein A. 2005. Sensitive genus-specific detection of Legionella by a 16S rRNA based sandwich hybridization assay. Journal of Microbiological Methods. 62(2):167-179. https://doi.org/10.1016/j.mimet.2005.02.008
3.
Rautio J, Barken K, Lahdenperä J, Breitenstein A, Molin S, Neubauer P. 2003. Microb Cell Fact. 2(1):4. https://doi.org/10.1186/1475-2859-2-4