Merck
HomeSmall Molecule HPLCMilli-Q® Advantage A10 for Water Purification
Milli-Q® Advantage A10 for Water Purification
An ultrapure water solution designed with you in mind
Overview
It's intelligent. It's intuitive. It's designed to make your work as pleasant and comfortable as possible — and to maximize lab productivity. Its breakthrough ergonomic design and powerful purification media ensure consistent production of superior-quality ultrapure water. See how the Milli-Q® IQ 7000 will change the way you work in the lab.
Features & Benefits at a Glance
  • Simple & intuitive dispensing: dispensing ultrapure water from the Q-POD® has become much easier, precise and effortless with the new “wheel” dispense. You can access a large number of dispensing functions that are useful for day-to-day work in the laboratory.
Milli-Q® IQ 7000 for Water Purification
  • Large, interactive touch screen: like a smartphone, the Q-POD® dispenser's interactive touch screen allows you to go anywhere in just a few clicks.
  • Optimized lab space: protect your working space from clutter! The system is designed for a neat installation and clean lab benches, free of loose tubing and cables. Just interact with your Q-POD® dispenser — the rest of the compact system can easily be integrated in the lab environment. For labs needing more than one ultrapure water point-of-use, up to four Q-POD® dispensers can be connected.
  • Superior ultrapure water — consistently — to the trace level: the Milli-Q® IQ 7000 system provides superior ultrapure water consistently to the trace level with the combination of Jetpore® and IQnano® resin. You can fully rely on the system's water purity for your critical experiments. And with the A10® TOC on-board monitoring, you're spared from uncertainty or misinterpreting results — and also from having to repeat time-consuming and costly analyses.
  • Improved sustainability: our products are designed with a focus on sustainability. The Milli-Q® IQ 7000 system uses mercury-free, environmentally friendly UV oxidation lamps. Additionally, the “LabClose” mode puts the system into "hibernation," to save energy while preserving the system water quality when you are away from the lab.
  • Advanced traceability: advanced data tracking is an integral part of Milli-Q® IQ 7000. Dispensing Events ensures a history of all dispense reports, and Daily Quality Measures provides water quality traceability over time. Complete History is a global archive of all system data when more in-depth analysis is required. Traceability is also ensured through the e-Sure tags on system POD-Paks and IPAK cartridges.
  • Quality & Compliance: its validated and qualified design, combined with our strict manufacturing Quality Assurance system, is your best warranty. We offer a complete range of services, including validation support, which is provided by certified field service engineers using calibrated equipment and a Milli-Q®-specific Qualification Workbook. A report on Milli-Q® water conformity to Type 1 Water quality as described by ASTM®, ISO® 3696 and CLSI® norms, and to Purified Water as described in USP and EP, is available upon request.
Video: Invest in Lab Efficiency
See how the Milli-Q® IQ 7000 can change your daily work in the lab with its ergonomic design, precise dispensing options, high-definition digital touch screen, integrated data management system, effortless maintenance, and much more.
Other Milli-Q® IQ 7000 Videos
  • A Touch of Intelligence
  • A Touch of Precision
  • A Touch of Efficiency
Water Specifications / International Conformity
Ultrapure, Type 1 water specifications1 (from Q-POD® dispenser)
1 These values are typical and may vary depending on the nature and concentration of contaminants in the feed water.
2 Resistivity can also be displayed non-temperature-compensated as required by USP.
3 In the appropriate operating conditions, otherwise typically ≤ 5ppb.
4 With Millipak® and Millipak® Gold
5 With Biopak®
6 With Millipak® Gold installed and used in a laminar flow hood
International Regulation Requirements
Declaration of CE and cUL Conformity:
The Milli-Q® IQ 7000 system has been tested by an independent and accredited company for compliance with CE directives related to safety and electromagnetic compatibility. The report can be consulted on demand. In addition, the Milli-Q® IQ 7000 system is built using components and practices recommended by UL and has been cUL marked. The registration can be verified on the UL web site: www.ul.com.