Skip to Content
Merck
Lithium cobalt(III) oxide 99.8% trace metals basis

Lithium cobalt oxide

Lithium cobalt(III) oxide 99.8% trace metals basis
Synonym(s):
Lithium cobaltite, Lithium cobalt(III) oxide
Linear Formula:
LiCoO2
Molecular Weight:
97.87

Products

Shipping
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2