Skip to Content
CL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts
Products
Product Category
cell lines (7)
stem cells (3)
media (1)
Brand
Sigma-Aldrich (11)
Biological Source
Quality Segment
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
PMEF-CFX
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
PMEF, Strain CF1, Irradiated, Passage 3
Expand
View Pricing
PMEF-DR4X-M
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Mouse
Expand
View Pricing
PMEF-DR4-M
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Mouse
Expand
View Pricing
PMEF-CF
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Expand
View Pricing
PMEF-N
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Expand
View Pricing
PMEF-CFL
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Expand
View Pricing
PMEF-CFL-P1
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Mouse
Expand
View Pricing
PMEF-HL
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Mouse
Expand
View Pricing
PMEF-NL
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Expand
View Pricing
PMEF-NX
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
Mouse
Expand
View Pricing
PMEF-NL-P1
EmbryoMax
®
Primary Mouse Embryonic Fibroblasts,
PMEF, Neo Resistant, Strain FVB, Not Treated, Passage 1
Expand
View Pricing