MilliporeSigma
  • Mechanisms underlying platelet function defect in a pedigree with familial platelet disorder with a predisposition to acute myelogenous leukemia: potential role for candidate RUNX1 targets.
Mechanisms underlying platelet function defect in a pedigree with familial platelet disorder with a predisposition to acute myelogenous leukemia: potential role for candidate RUNX1 targets.
Journal of thrombosis and haemostasis : JTH (2014-03-13)
A C Glembotsky, D Bluteau, Y R Espasandin, N P Goette, R F Marta, C P Marin Oyarzun, L Korin, P R Lev, R P Laguens, F C Molinas, H Raslova, P G Heller
RÉSUMÉ
Familial platelet disorder with a predisposition to acute myelogenous leukemia (FPD/AML) is an inherited platelet disorder caused by a germline RUNX1 mutation and characterized by thrombocytopenia, a platelet function defect, and leukemia predisposition. The mechanisms underlying FPD/AML platelet dysfunction remain incompletely clarified. We aimed to determine the contribution of platelet structural abnormalities and defective activation pathways to the platelet phenotype. In addition, by using a candidate gene approach, we sought to identify potential RUNX1-regulated genes involved in these defects. Lumiaggregometry, α-granule and dense granule content and release, platelet ultrastructure, αIIb β3 integrin activation and outside-in signaling were assessed in members of one FPD/AML pedigree. Expression levels of candidate genes were measured and luciferase reporter assays and chromatin immunoprecipitation were performed to study NF-E2 regulation by RUNX1. A severe decrease in platelet aggregation, defective αIIb β3 integrin activation and combined αδ storage pool deficiency were found. However, whereas the number of dense granules was markedly reduced, α-granule content was heterogeneous. A trend towards decreased platelet spreading was found, and β3 integrin phosphorylation was impaired, reflecting altered outside-in signaling. A decrease in the level of transcription factor p45 NF-E2 was shown in platelet RNA and lysates, and other deregulated genes included RAB27B and MYL9. RUNX1 was shown to bind to the NF-E2 promoter in primary megakaryocytes, and wild-type RUNX1, but not FPD/AML mutants, was able to activate NF-E2 expression. The FPD/AML platelet function defect represents a complex trait, and RUNX1 orchestrates platelet function by regulating diverse aspects of this process. This study highlights the RUNX1 target NF-E2 as part of the molecular network by which RUNX1 regulates platelet biogenesis and function.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Chlorure de sodium, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Chlorure de magnésium solution, 1.00 M±0.01 M
Sigma-Aldrich
Chlorure de magnésium, ≥98%
Sigma-Aldrich
Chlorure de sodium solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Chlorure de sodium, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Chlorure de sodium solution, 5 M in H2O, BioReagent, suitable for cell culture
SAFC
Chlorure de sodium solution, 5 M
Sigma-Aldrich
Chlorure de magnésium, powder, <200 μm
Sigma-Aldrich
Chlorure de sodium solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Chlorure de sodium, 99.999% trace metals basis
Sigma-Aldrich
Chlorure de sodium, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Chlorure de magnésium solution, BioUltra, 2 M in H2O
Sigma-Aldrich
Chlorure de magnésium, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Chlorure de magnésium solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Chlorure de magnésium, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Chlorure de sodium, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Chlorure de sodium, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Chlorure de sodium, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Chlorure de sodium, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Chlorure de magnésium, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Chlorure de sodium solution, 5 M
Sigma-Aldrich
Chlorure de sodium, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chlorure de magnésium solution, 0.1 M
Sigma-Aldrich
Chlorure de sodium solution, 0.85%
Sigma-Aldrich
PAC-1, ≥98% (HPLC)
Supelco
Chlorure de sodium, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Chlorure de magnésium solution, BioUltra, ~1 M in H2O
Supelco
Chlorure de sodium, Pharmaceutical Secondary Standard; Certified Reference Material