Principaux documents
TMO140067
Nunc® EasyFill Cell Factory filter assembly cap
high-density polyethylene, pore size 1.0 μm
Synonyme(s) :
closed cell system cap, closed cell system filter cap
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.13
Matériaux
high-density polyethylene
Fabricant/nom de marque
Nunc 140067
Dimension de pores
1.0 μm pore size
Description générale
Further the development of applications that require a more closed cell culture system using Thermo Scientific Nunc Cell Factory Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries