MilliporeSigma
  • Airway smooth muscle cells synthesize hyaluronan cable structures independent of inter-alpha-inhibitor heavy chain attachment.
Airway smooth muscle cells synthesize hyaluronan cable structures independent of inter-alpha-inhibitor heavy chain attachment.
The Journal of biological chemistry (2008-12-17)
Mark E Lauer, Csaba Fulop, Durba Mukhopadhyay, Suzy Comhair, Serpil C Erzurum, Vincent C Hascall
ABSTRACT
The covalent association of inter-alpha-inhibitor-derived heavy chains (HCs) with hyaluronan was first described in synovial fluid from arthritic patients and later described as a structural and functional component of hyaluronan "cable" structures produced by many different cells and stimuli. HC transfer has been shown to be mediated by the protein product of TSG-6 (tumor necrosis factor-stimulated gene 6). Considering the accumulation of hyaluronan in airways following asthmatic attacks and the subsequent infiltration of leukocytes, we sought to characterize HC substitution of hyaluronan "cables" in primary mouse airway smooth muscle cells (MASM) and primary human airway smooth muscle cells (HASM). We found that cells derived from mice lacking TSG-6 had no defect in hyaluronan production or hyaluronan-mediated leukocyte adhesion when treated with the viral mimic poly(I,C). Functional hyaluronan cables were induced by cycloheximide in the confirmed absence of protein synthesis, with or without simultaneous treatment with poly(I,C). We characterized the species specificity of the antibody other investigators used to describe the HC-hyaluronan complex of hyaluronan cables and found minimal affinity to bovine-derived HCs in contrast to HCs from mouse and human sera. Thus, we cultured MASM and HASM cells in serum from these three sources and analyzed hyaluronan extracts for HCs and other hyaluronan-binding proteins, using parallel cumulus cell-oocyte complex (COC) extracts as positive controls. We conclude that, if hyaluronan cables derived from MASM and HASM cells are substituted with HCs, the amount of substitution is significantly below the limit of detection when compared with COC extracts of similar hyaluronan mass.
Sigma-Aldrich
Sera from mouse, frozen liquid (from clotted whole blood)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Versican Antibody, a.a. 535-598 of mouse versican, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Versican Antibody, a.a. 1360-1439 of mouse versican, Chemicon®, from rabbit
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, γ-irradiated