MilliporeSigma
Bottlenose dolphins (
International journal for parasitology. Parasites and wildlife (2017-09-28)
R Villagra-Blanco, L M R Silva, A Aguilella-Segura, I Arcenillas-Hernández, C Martínez-Carrasco, A Seipp, U Gärtner, R Ruiz de Ybañez, A Taubert, C Hermosilla
ABSTRACT
Neutrophil extracellular traps (NETs) are web-like structures composed of nuclear DNA decorated with histones and cytoplasmic peptides which antiparasitic properties have not previously been investigated in cetaceans. Polymorphonuclear neutrophils (PMN) were isolated from healthy bottlenose dolphins (
Sigma-Aldrich
Anti-Histone Antibody, clone H11-4, clone H11-4, Chemicon®, from mouse
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%