MilliporeSigma
  • Issues with the Specificity of Immunological Reagents for NLRP3: Implications for Age-related Macular Degeneration.
Issues with the Specificity of Immunological Reagents for NLRP3: Implications for Age-related Macular Degeneration.
Scientific reports (2018-01-13)
Cassandra Kosmidou, Nikolaos E Efstathiou, Mien V Hoang, Shoji Notomi, Eleni K Konstantinou, Masayuki Hirano, Kosuke Takahashi, Daniel E Maidana, Pavlina Tsoka, Lucy Young, Evangelos S Gragoudas, Timothy W Olsen, Yuki Morizane, Joan W Miller, Demetrios G Vavvas
ABSTRACT
Contradictory data have been presented regarding the implication of the NACHT, LRR and PYD domains-containing protein 3 (NLRP3) inflammasome in age-related macular degeneration (AMD), the leading cause of vision loss in the Western world. Recognizing that antibody specificity may explain this discrepancy and in line with recent National Institutes of Health (NIH) guidelines requiring authentication of key biological resources, the specificity of anti-NLRP3 antibodies was assessed to elucidate whether non-immune RPE cells express NLRP3. Using validated resources, NLRP3 was not detected in human primary or human established RPE cell lines under multiple inflammasome-priming conditions, including purported NLRP3 stimuli in RPE such as DICER1 deletion and Alu RNA transfection. Furthermore, NLRP3 was below detection limits in ex vivo macular RPE from AMD patients, as well as in human induced pluripotent stem cell (hiPSC)-derived RPE from patients with overactive NLRP3 syndrome (Chronic infantile neurologic cutaneous and articulate, CINCA syndrome). Evidence presented in this study provides new data regarding the interpretation of published results reporting NLRP3 expression and upregulation in RPE and addresses the role that this inflammasome plays in AMD pathogenesis.
Sigma-Aldrich
Interleukin-1α human, IL-1α, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-NLRP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Brilliant Blue G - Colloidal Concentrate