MilliporeSigma
  • The WNT target SP5 negatively regulates WNT transcriptional programs in human pluripotent stem cells.
The WNT target SP5 negatively regulates WNT transcriptional programs in human pluripotent stem cells.
Nature communications (2017-10-19)
Ian J Huggins, Tomas Bos, Olivia Gaylord, Christina Jessen, Brianna Lonquich, Angeline Puranen, Jenna Richter, Charlotte Rossdam, David Brafman, Terry Gaasterland, Karl Willert
ABSTRACT
The WNT/β-catenin signaling pathway is a prominent player in many developmental processes, including gastrulation, anterior-posterior axis specification, organ and tissue development, and homeostasis. Here, we use human pluripotent stem cells (hPSCs) to study the dynamics of the transcriptional response to exogenous activation of the WNT pathway. We describe a mechanism involving the WNT target gene SP5 that leads to termination of the transcriptional program initiated by WNT signaling. Integration of gene expression profiles of wild-type and SP5 mutant cells with genome-wide SP5 binding events reveals that SP5 acts to diminish expression of genes previously activated by the WNT pathway. Furthermore, we show that activation of SP5 by WNT signaling is most robust in cells with developmental potential, such as stem cells. These findings indicate a mechanism by which the developmental WNT signaling pathway reins in expression of transcriptional programs.
Sigma-Aldrich
Anti-Dimethyl Histone H3 (Lys4) Antibody, clone CMA303, clone CMA303, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, culture supernatant, clone MC315, Upstate®
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid