MilliporeSigma
  • Regulatory effects of simvastatin and apoJ on APP processing and amyloid-β clearance in blood-brain barrier endothelial cells.
Regulatory effects of simvastatin and apoJ on APP processing and amyloid-β clearance in blood-brain barrier endothelial cells.
Biochimica et biophysica acta (2017-09-25)
Martina Zandl-Lang, Elham Fanaee-Danesh, Yidan Sun, Nicole M Albrecher, Chaitanya Chakravarthi Gali, Igor Čančar, Alexandra Kober, Carmen Tam-Amersdorfer, Anika Stracke, Steffen M Storck, Ahmed Saeed, Jasminka Stefulj, Claus U Pietrzik, Mark R Wilson, Ingemar Björkhem, Ute Panzenboeck
ABSTRACT
Amyloid-β peptides (Aβ) accumulate in cerebral capillaries indicating a central role of the blood-brain barrier (BBB) in the pathogenesis of Alzheimer's disease (AD). Although a relationship between apolipoprotein-, cholesterol- and Aβ metabolism is evident, the interconnecting mechanisms operating in brain capillary endothelial cells (BCEC) are poorly understood. ApoJ (clusterin) is present in HDL that regulates cholesterol metabolism which is disturbed in AD. ApoJ levels are increased in AD brains and in plasma of cerebral amyloid angiopathy (CAA) patients. ApoJ may bind, prevent fibrillization, and enhance clearance of Aβ. We here define a connection of apoJ and cellular cholesterol homeostasis in amyloid precursor protein (APP) processing/Aβ metabolism at the BBB. Silencing of apoJ in primary porcine (p)BCEC decreased intracellular APP and Aβ oligomer levels while the addition of purified apoJ to pBCEC increased intracellular APP and enhanced Aβ clearance across the pBCEC monolayer. Treatment of pBCEC with Aβ
Sigma-Aldrich
Anti-Amyloid Oligomer Antibody, αβ, oligomeric, serum, Chemicon®
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
24(S)-Hydroxycholesterol, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution