MilliporeSigma
MitoNeoD: A Mitochondria-Targeted Superoxide Probe.
Cell chemical biology (2017-09-12)
Maria M Shchepinova, Andrew G Cairns, Tracy A Prime, Angela Logan, Andrew M James, Andrew R Hall, Sara Vidoni, Sabine Arndt, Stuart T Caldwell, Hiran A Prag, Victoria R Pell, Thomas Krieg, John F Mulvey, Pooja Yadav, James N Cobley, Thomas P Bright, Hans M Senn, Robert F Anderson, Michael P Murphy, Richard C Hartley
ABSTRACT
Mitochondrial superoxide (O2⋅-) underlies much oxidative damage and redox signaling. Fluorescent probes can detect O2⋅-, but are of limited applicability in vivo, while in cells their usefulness is constrained by side reactions and DNA intercalation. To overcome these limitations, we developed a dual-purpose mitochondrial O2⋅- probe, MitoNeoD, which can assess O2⋅- changes in vivo by mass spectrometry and in vitro by fluorescence. MitoNeoD comprises a O2⋅--sensitive reduced phenanthridinium moiety modified to prevent DNA intercalation, as well as a carbon-deuterium bond to enhance its selectivity for O2⋅- over non-specific oxidation, and a triphenylphosphonium lipophilic cation moiety leading to the rapid accumulation within mitochondria. We demonstrated that MitoNeoD was a versatile and robust probe to assess changes in mitochondrial O2⋅- from isolated mitochondria to animal models, thus offering a way to examine the many roles of mitochondrial O2⋅- production in health and disease.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Sigma-Aldrich
MitoPQ, ≥90% (HPLC)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder