MilliporeSigma
  • The silanol content and in vitro cytolytic activity of flame-made silica.
The silanol content and in vitro cytolytic activity of flame-made silica.
Journal of colloid and interface science (2017-08-07)
Anastasia Spyrogianni, Inge K Herrmann, Kerda Keevend, Sotiris E Pratsinis, Karsten Wegner
ABSTRACT
The surface chemistry of synthetic amorphous silicas is essential for their applicational performance and for understanding their interactions with biological matter. Synthesis of silica by flame spray pyrolysis (FSP) allows to control the content and type of hydroxyl groups which also affects the cytolytic activity. By controlling the FSP process variables, silica nanoparticles with the same specific surface area but different surface chemistry and content of internal silanols are prepared by combustion of hexamethyldisiloxane sprays, as characterized by Raman and infrared spectroscopy, thermogravimetric analysis, and titration with lithium alanate. Cytolytic activity is assessed in terms of membrane damage in human blood monocytes in vitro. Unlike commercial fumed silica, FSP-made silicas contain a significant amount of internal silanol groups and a high surface hydroxyl density, up to ∼8OH/nm
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)