MilliporeSigma
LRRK2 promotes the activation of NLRC4 inflammasome during
The Journal of experimental medicine (2017-08-20)
Weiwei Liu, Xia'nan Liu, Yu Li, Junjie Zhao, Zhenshan Liu, Zhuqin Hu, Ying Wang, Yufeng Yao, Aaron W Miller, Bing Su, Mark R Cookson, Xiaoxia Li, Zizhen Kang
ABSTRACT
Although genetic polymorphisms in the