MilliporeSigma
  • A novel requirement for DROSHA in maintenance of mammalian CG methylation.
A novel requirement for DROSHA in maintenance of mammalian CG methylation.
Nucleic acids research (2017-09-22)
Athanasia Stathopoulou, Jyoti B Chhetri, John C Ambrose, Pierre-Olivier Estève, Lexiang Ji, Hediye Erdjument-Bromage, Guoqiang Zhang, Thomas A Neubert, Sriharsa Pradhan, Javier Herrero, Robert J Schmitz, Steen K T Ooi
ABSTRACT
In mammals, faithful inheritance of genomic methylation patterns ensures proper gene regulation and cell behaviour, impacting normal development and fertility. Following establishment, genomic methylation patterns are transmitted through S-phase by the maintenance methyltransferase Dnmt1. Using a protein interaction screen, we identify Microprocessor component DROSHA as a novel DNMT1-interactor. Drosha-deficient embryonic stem (ES) cells display genomic hypomethylation that is not accounted for by changes in the levels of DNMT proteins. DNMT1-mediated methyltransferase activity is also reduced in these cells. We identify two transcripts that are specifically upregulated in Drosha- but not Dicer-deficient ES cells. Regions within these transcripts predicted to form stem-loop structures are processed by Microprocessor and can inhibit DNMT1-mediated methylation in vitro. Our results highlight DROSHA as a novel regulator of mammalian DNA methylation and we propose that DROSHA-mediated processing of RNA is necessary to ensure full DNMT1 activity. This adds to the DROSHA repertoire of non-miRNA dependent functions as well as implicating RNA in regulating DNMT1 activity and correct levels of genomic methylation.
Sigma-Aldrich
DNMT1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥50% (SDS-PAGE)
Sigma-Aldrich
Anti-Tet2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture