MilliporeSigma
  • Neutrophil polarization by IL-27 as a therapeutic target for intracerebral hemorrhage.
Neutrophil polarization by IL-27 as a therapeutic target for intracerebral hemorrhage.
Nature communications (2017-09-21)
Xiurong Zhao, Shun-Ming Ting, Chin-Hsuan Liu, Guanghua Sun, Marian Kruzel, Meaghan Roy-O'Reilly, Jaroslaw Aronowski
ABSTRACT
Shortly after intracerebral hemorrhage, neutrophils infiltrate the intracerebral hemorrhage-injured brain. Once within the brain, neutrophils degranulate, releasing destructive molecules that may exacerbate brain damage. However, neutrophils also release beneficial molecules, including iron-scavenging lactoferrin that may limit hematoma/iron-mediated brain injury after intracerebral hemorrhage. Here, we show that the immunoregulatory cytokine interleukin-27 is upregulated centrally and peripherally after intracerebral hemorrhage. Data from rodent models indicate that interleukin-27 modifies neutrophil maturation in the bone marrow, suppressing their production of pro-inflammatory/cytotoxic products while increasing their production of beneficial iron-scavenging molecules, including lactoferrin. Finally, interleukin-27 or lactoferrin administration results in reduced edema, enhanced hematoma clearance, and improved neurological outcomes in an animal model of intracerebral hemorrhage. These results suggest that interleukin-27/lactoferrin-mediated modulations of neutrophil function may represent a therapeutically viable concept for the modification of neutrophils toward a "beneficial" phenotype for the treatment of intracerebral hemorrhage.Neutrophils are important modulators of tissue damage after intracerebral hemorrhage (ICH), but how this function is regulated is not clear. Here, the authors show interleukin-27 promotes the tissue-protecting functions of neutrophils via, at least partly, the induction of lactoferrin to present a potential therapy for ICH.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Millipore
MILLIPLEX® Mouse TH17 Magnetic Bead Panel - Premixed 25 plex - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Sigma-Aldrich
Human LTF / Lactotransferrin ELISA Kit