MilliporeSigma
  • Vitamin D status in growing dairy goats and sheep: Influence of ultraviolet B radiation on bone metabolism and calcium homeostasis.
Vitamin D status in growing dairy goats and sheep: Influence of ultraviolet B radiation on bone metabolism and calcium homeostasis.
Journal of dairy science (2017-08-22)
M V Nemeth, M R Wilkens, A Liesegang
ABSTRACT
The aim of this study was to investigate how controlled UVB irradiation in combination with reduced nutritional vitamin D (vitD) supply affects vitD status and Ca metabolism of growing goats and sheep. The hypothesis was that, like dairy cows, goats and sheep are able to compensate for the missing nutritional supply of vitD through endogenous production in the skin, with the consequence of a high vitD status and a balanced Ca homeostasis. Sixteen lambs and 14 goat kids aged 3 and a half months were housed in an UVB free environment and fed hay and a vitD-free concentrate over a period of 13 wk. One group of each species was exposed to UVB lamps daily during individual feeding; the other groups served as controls. Serum, urine, and feces samples were taken at the start and at a monthly interval. Serum was analyzed for vitD metabolites, bone markers, growth hormone, insulin-like growth factor I, Ca, and P. Apparent digestibility and urinary excretion of Ca and P were determined. The left metatarsus was analyzed by peripheral quantitative computer tomography for bone mineral density before starting and at the end of the trial. In wk 13, all animals were slaughtered and samples of skin, rumen, duodenum, kidney, and bone (metatarsus) were collected. Content of sterols of vitD synthesis in the skin, Ca flux rates in rumen and duodenum, expression of vitD receptor in duodenum and kidney, renal and intestinal gene expression of Ca transport proteins, and renal enzymes related to vitD metabolism were determined. The UVB exposure led to lower 7-dehydrocholesterol content in the skin and a better vitD status (higher serum 25-hydroxyvitamin D), but no signs of vitD deficiency were seen in the control groups and no effect of irradiation was detected in the analyzed parameters of Ca homeostasis. Differences between the 2 species were detected: lambs had a higher increase of bone mineral density, lower values of bone markers, growth hormone, and insulin-like growth factor I in serum and higher tachysterol and lower lumisterol content in skin compared with goat kids. The results indicated that growing lambs and goat kids are able to compensate for a vitD-reduced diet by cutaneous vitD synthesis when exposed to UVB irradiation and therefore to keep a high vitD status. In contrast, when a reduced vitD diet is combined with missing UVB exposure, the vitD status drops, but the experimental time was probably too short to induce a vitD deficiency or an effect on Ca homeostasis.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Human GH ELISA Kit, for serum, plasma, cell culture supernatant and urine