MilliporeSigma
  • Autophagy is required for cell survival under L-asparaginase-induced metabolic stress in acute lymphoblastic leukemia cells.
Autophagy is required for cell survival under L-asparaginase-induced metabolic stress in acute lymphoblastic leukemia cells.
Oncogene (2017-03-28)
H Takahashi, J Inoue, K Sakaguchi, M Takagi, S Mizutani, J Inazawa
ABSTRACT
L-asparaginase has been used for more than three decades in acute lymphoblastic leukemia (ALL) patients and remains an essential drug in the treatment of ALL. Poor response to L-asparaginase is associated with increased risk of therapeutic failure in ALL. However, both the metabolic perturbation and molecular context of L-asparaginase-treated ALL cells has not been fully elucidated. Here we identify that treatment with L-asparaginase results in metabolic shutdown via the reduction of both glycolysis and oxidative phosphorylation, accompanied by mitochondrial damage and activation of autophagy. The autophagy is involved in reducing reactive oxygen species (ROS) level by eliminating injured mitochondria. Inhibition of autophagy enhances L-asparaginase-induced cytotoxicity and overcomes the acquired resistance to L-asparaginase in ALL cells. The ROS-p53-positive feedback loop is an essential mechanism of this synergistic cytotoxicity. Thus, our findings provide the rationale for the future development of combined treatment of L-asparaginase and anti-autophagy drug in ALL patients.
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), lyophilized, clone DO-1, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Sigma-Aldrich
Triton X-100, laboratory grade