MilliporeSigma
  • Brain-derived neurotrophic factor of the cerebral microvasculature: a forgotten and nitric oxide-dependent contributor of brain-derived neurotrophic factor in the brain.
Brain-derived neurotrophic factor of the cerebral microvasculature: a forgotten and nitric oxide-dependent contributor of brain-derived neurotrophic factor in the brain.
Acta physiologica (Oxford, England) (2016-07-02)
A Monnier, A Prigent-Tessier, A Quirié, N Bertrand, S Savary, C Gondcaille, P Garnier, C Demougeot, C Marie
ABSTRACT
Evidence that brain-derived neurotrophic factor (BDNF), a neurotrophin largely involved in cognition, is expressed by cerebral endothelial cells led us to explore in rats the contribution of the cerebral microvasculature to BDNF found in brain tissue and the link between cerebrovascular nitric oxide (NO) and BDNF production. Brain BDNF protein levels were measured before and after in situ removal of the cerebral endothelium that was achieved by brain perfusion with a 0.2% CHAPS (3-[(3-cholamidopropyl) dimethylammonio]-1-propane sulphonate) solution. BDNF protein and mRNA levels as well as levels of endothelial NO synthase phosphorylated at serine 1177 (P-eNOS CHAPS perfusion resulted in a marked decrease in brain BDNF levels. Hypertension decreased and exercise increased P-eNOS These data reveal that BDNF levels measured in brain homogenates correspond for a large part to BDNF present in cerebral endothelial cells and that cerebrovascular BDNF production is dependent on cerebrovascular endothelial eNOS activity. They provide a paradigm shift in the cellular source of brain BDNF and suggest a new approach to improve our understanding of the link between endothelial function and cognition.
Sigma-Aldrich
Anti-GLUT1 (CT) Antibody, clone 5B12.3, clone 5B12.3, from mouse
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-von Willebrand Factor antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
CHAPS hydrate, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse