MilliporeSigma
  • LY3009120, a panRAF inhibitor, has significant anti-tumor activity in BRAF and KRAS mutant preclinical models of colorectal cancer.
LY3009120, a panRAF inhibitor, has significant anti-tumor activity in BRAF and KRAS mutant preclinical models of colorectal cancer.
Oncotarget (2016-12-22)
Eliza Vakana, Susan Pratt, Wayne Blosser, Michele Dowless, Nicholas Simpson, Xiu-Juan Yuan, Susan Jaken, Jason Manro, Jennifer Stephens, Youyan Zhang, Lysiane Huber, Sheng-Bin Peng, Louis F Stancato
ABSTRACT
Activating mutations in the KRAS and BRAF genes, leading to hyperactivation of the RAS/RAF/MAPK oncogenic signaling cascade, are common in patients with colorectal cancer (CRC). While selective BRAF inhibitors are efficacious in BRAFmut melanoma, they have limited efficacy in BRAFmut CRC patients. In a RASmut background, selective BRAF inhibitors are contraindicated due to paradoxical activation of the MAPK pathway through potentiation of CRAF kinase activity. A way to overcome such paradoxical activation is through concurrent inhibition of the kinase activity of both RAF isoforms. Here, we further examined the effects of LY3009120, a panRAF and RAF dimer inhibitor, in human models of CRC with various mutational backgrounds. We demonstrate that LY3009120 induced anti-proliferative effects in BRAFmut and KRASmut CRC cell lines through G1-cell cycle arrest. The anti-proliferative effects of LY3009120 in KRASmut CRC cell lines phenocopied molecular inhibition of RAF isoforms by simultaneous siRNA-mediated knockdown of ARAF, BRAF and CRAF. Additionally, LY3009120 displayed significant activity in in vivo BRAFmut and KRASmut CRC xenograft models. Examination of potential resistance to LY3009120 demonstrated RAF-independent ERK and AKT activation in the KRASmut CRC cell line HCT 116. These findings describe the preclinical activity of a panRAF inhibitor in a BRAFmut and KRASmut CRC setting.
Sigma-Aldrich
LY3009120, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade