MilliporeSigma
  • Short-interfering RNA-mediated Lck knockdown results in augmented downstream T cell responses.
Short-interfering RNA-mediated Lck knockdown results in augmented downstream T cell responses.
Journal of immunology (Baltimore, Md. : 1950) (2005-11-23)
Trond Methi, Jacob Ngai, Milada Mahic, Mohammed Amarzguioui, Torkel Vang, Kjetil Tasken
ABSTRACT
The Src family kinase Lck is essential for T cell Ag receptor-mediated signaling. In this study, we report the effects of acute elimination of Lck in Jurkat TAg and primary T cells using RNA interference mediated by short-interfering RNAs. In cells with Lck knockdown (kd), proximal TCR signaling was strongly suppressed as indicated by reduced zeta-chain phosphorylation and intracellular calcium mobilization. However, we observed sustained and elevated phosphorylation of ERK1/2 in Lck kd cells 30 min to 2 h after stimulation. Downstream effects on immune function as determined by activation of a NFAT-AP-1 reporter, and TCR/CD28-stimulated IL-2 secretion were strongly augmented in Jurkat and primary T cells, respectively. As expected, overexpression of SHP-1 in Jurkat cells inhibited TCR-induced NFAT-AP-1 activation, but this effect could be overcome by simultaneous kd of Lck. Furthermore, acute elimination of Lck also suppressed TCR-mediated activation of SHP-1, suggesting the possible role of SHP-1 in a negative feedback loop originating from Lck. This report underscores Lck as an important mediator of proximal TCR signaling, but also indicates a suppressive role on downstream immune function.
Sigma-Aldrich
Anti-LAT Antibody, Upstate®, from rabbit
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder