MilliporeSigma
  • Calorie restriction protects against experimental abdominal aortic aneurysms in mice.
Calorie restriction protects against experimental abdominal aortic aneurysms in mice.
The Journal of experimental medicine (2016-11-05)
Yue Liu, Ting-Ting Wang, Ran Zhang, Wen-Yan Fu, Xu Wang, Fang Wang, Peng Gao, Yang-Nan Ding, Yan Xie, De-Long Hao, Hou-Zao Chen, De-Pei Liu
ABSTRACT
Abdominal aortic aneurysm (AAA), characterized by a localized dilation of the abdominal aorta, is a life-threatening vascular pathology. Because of the current lack of effective treatment for AAA rupture, prevention is of prime importance for AAA management. Calorie restriction (CR) is a nonpharmacological intervention that delays the aging process and provides various health benefits. However, whether CR prevents AAA formation remains untested. In this study, we subjected Apoe
Sigma-Aldrich
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, clone 62-141-13, Upstate®, from rabbit
Sigma-Aldrich
SIRT1 Assay Kit, sufficient for 100 assays
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid