MilliporeSigma
  • ROCK- and myosin-dependent matrix deformation enables protease-independent tumor-cell invasion in vivo.
ROCK- and myosin-dependent matrix deformation enables protease-independent tumor-cell invasion in vivo.
Current biology : CB (2006-08-08)
Jeffrey B Wyckoff, Sophie E Pinner, Steve Gschmeissner, John S Condeelis, Erik Sahai
ABSTRACT
Tumor cells invading three-dimensional matrices need to remodel the extracellular matrix (ECM) in their path. Many studies have focused on the role of extracellular proteases; however, cells with amoeboid or rounded morphologies are able to invade even when these enzymes are inhibited. Here, we describe the mechanism by which cells move through a dense ECM without proteolysis. Amoeboid tumor cells generate sufficient actomyosin force to deform collagen fibers and are able to push through the ECM. Force generation is elevated in metastatic MTLn3E cells, and this correlates with increased invasion and altered myosin light chain (MLC) organization. In metastatic cells, MLC is organized perpendicularly to the direction of movement behind the invading edge. Both the organization of MLC and force generation are dependent upon ROCK function. We demonstrate that ROCK regulates the phosphorylation of MLC just behind the invading margin of the cell. Imaging of live tumors shows that MLC is organized in a similar ROCK-dependent fashion in vivo and that inhibition of ROCK but not matrix-metalloproteases reduces cancer cell motility in vivo.
Sigma-Aldrich
Rho Kinase Inhibitor, The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).
Sigma-Aldrich
(-)-Blebbistatin, The active enantiomer of (±)-Blebbistatin that accounts for the inhibitory activity towards ATPase and myosin II-dependent cellular processes.
Sigma-Aldrich
Monoclonal Anti-Myosin Light Chain Kinase antibody produced in mouse, clone K36, ascites fluid
Sigma-Aldrich
GM 6001, GM 6001, CAS 142880-36-2, is a potent, cell-permeable, broad-spectrum inhibitor of matrix metalloproteinases.