MilliporeSigma
  • Lactobacillus casei improves resistance to pneumococcal respiratory infection in malnourished mice.
Lactobacillus casei improves resistance to pneumococcal respiratory infection in malnourished mice.
The Journal of nutrition (2005-06-03)
Julio Villena, Silvia Racedo, Graciela Agüero, Elena Bru, Marcela Medina, Susana Alvarez
ABSTRACT
We studied the effect of Lactobacillus casei CRL 431 used as a supplement in a repletion diet on the resistance to Streptococcus pneumoniae respiratory infection in malnourished mice. Weaned mice were malnourished after they consumed a protein-free diet (PFD) for 21 d. Malnourished mice were fed a balanced conventional diet (BCD) with or without supplemental L. casei for 7, 14, or 21 consecutive days, or BCD for 7 d with L. casei supplementation on d 6 and 7 (7dBCD+2dLc). The malnourished control (MNC) group was fed only the PFD, whereas well-nourished control (WNC) mice consumed the BCD ad libitum. Mice were challenged with S. pneumoniae at the end of each dietary treatment. Lung colonization and bacteremia were significantly greater in MNC than in WNC. Normalization of the immune response occurred in malnourished mice fed the BCD for 21 d. L. casei supplementation reduced the time required for a normal response from 21 to 7 d. Mice administered the 7dBCD+2dLc repletion treatment had a more effective pathogen clearance from blood and significantly lower lung damage than MNC. This treatment improved both the number of leukocytes and neutrophils in blood and bronchoalveolar lavages (BAL) and the bactericidal function of phagocytic cells to levels that did not differ from those of WNC. In the 7dBCD+2dLc mice, antipneumococcal IgA in BAL was higher than in WNC, whereas antipneumococcal IgG in serum and BAL did not differ. This study suggests that the addition of L. casei to the repletion diet has a beneficial effect because it accelerates the recovery of the innate immune response and improves the specific immune mechanisms against an S. pneumoniae respiratory infection in malnourished mice.
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
IgA, Kappa from murine myeloma, clone TEPC 15, purified immunoglobulin, buffered aqueous solution