MilliporeSigma
  • Oxidative stress contributes to aging by enhancing pancreatic angiogenesis and insulin signaling.
Oxidative stress contributes to aging by enhancing pancreatic angiogenesis and insulin signaling.
Cell metabolism (2008-02-06)
Gaëlle Laurent, Florence Solari, Bogdan Mateescu, Melis Karaca, Julien Castel, Brigitte Bourachot, Christophe Magnan, Marc Billaud, Fatima Mechta-Grigoriou
ABSTRACT
JunD, a transcription factor of the AP-1 family, protects cells against oxidative stress. Here, we show that junD(-/-) mice exhibit features of premature aging and shortened life span. They also display persistent hypoglycemia due to enhanced insulin secretion. Consequently, the insulin/IGF-1 signaling pathways are constitutively stimulated, leading to inactivation of FoxO1, a positive regulator of longevity. Hyperinsulinemia most likely results from enhanced pancreatic islet vascularization owing to chronic oxidative stress. Indeed, accumulation of free radicals in beta cells enhances VEGF-A transcription, which in turn increases pancreatic angiogenesis and insulin secretion. Accordingly, long-term treatment with an antioxidant rescues the phenotype of junD(-/-) mice. Indeed, dietary antioxidant supplementation was protective against pancreatic angiogenesis, hyperinsulinemia, and subsequent activation of insulin signaling cascades in peripheral tissues. Taken together, these data establish a pivotal role for oxidative stress in systemic regulation of insulin and define a key role for the JunD protein in longevity.
Sigma-Aldrich
Anti-phospho-IR (Tyr972) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Galactosidase antibody produced in mouse, clone GAL-13, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid