MilliporeSigma
  • Acute heart failure with cardiomyocyte atrophy induced in adult mice by ablation of cardiac myosin light chain kinase.
Acute heart failure with cardiomyocyte atrophy induced in adult mice by ablation of cardiac myosin light chain kinase.
Cardiovascular research (2016-03-31)
Michael T Massengill, Hassan M Ashraf, Rajib R Chowdhury, Stephen M Chrzanowski, Jeena Kar, Sonisha A Warren, Glenn A Walter, Huadong Zeng, Byung-Ho Kang, Robert H Anderson, Richard L Moss, Hideko Kasahara
ABSTRACT
Under pressure overload, initial adaptive hypertrophy of the heart is followed by cardiomyocyte elongation, reduced contractile force, and failure. The mechanisms governing the transition to failure are not fully understood. Pressure overload reduced cardiac myosin light chain kinase (cMLCK) by ∼80% within 1 week and persists. Knockdown of cMLCK in cardiomyocytes resulted in reduced cardiac contractility and sarcomere disorganization. Thus, we hypothesized that acute reduction of cMLCK may be causative for reduced contractility and cardiomyocyte remodelling during the transition from compensated to decompensated cardiac hypertrophy. To mimic acute cMLCK reduction in adult hearts, the floxed-Mylk3 gene that encodes cMLCK was inducibly ablated in Mylk3(flox/flox)/merCremer mice (Mylk3-KO), and compared with two control mice (Mylk3(flox/flox) and Mylk3(+/+)/merCremer) following tamoxifen injection (50 mg/kg/day, 2 consecutive days). In Mylk3-KO mice, reduction of cMLCK protein was evident by 4 days, with a decline to below the level of detection by 6 days. By 7 days, these mice exhibited heart failure, with reduction of fractional shortening compared with those in two control groups (19.8 vs. 28.0% and 27.7%). Severely convoluted cardiomyocytes with sarcomeric disorganization, wavy fibres, and cell death were demonstrated in Mylk3-KO mice. The cardiomyocytes were also unable to thicken adaptively to pressure overload. Our results, using a new mouse model mimicking an acute reduction of cMLCK, suggest that cMLCK plays a pivotal role in the transition from compensated to decompensated hypertrophy via sarcomeric disorganization.
Sigma-Aldrich
Anti-phospho-Phospholamban (Ser16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Troponin T antibody produced in mouse, clone JLT-12, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse