MilliporeSigma
  • Identification of chemicals that mimic transcriptional changes associated with autism, brain aging and neurodegeneration.
Identification of chemicals that mimic transcriptional changes associated with autism, brain aging and neurodegeneration.
Nature communications (2016-04-01)
Brandon L Pearson, Jeremy M Simon, Eric S McCoy, Gabriela Salazar, Giulia Fragola, Mark J Zylka
ABSTRACT
Environmental factors, including pesticides, have been linked to autism and neurodegeneration risk using retrospective epidemiological studies. Here we sought to prospectively identify chemicals that share transcriptomic signatures with neurological disorders, by exposing mouse cortical neuron-enriched cultures to hundreds of chemicals commonly found in the environment and on food. We find that rotenone, a pesticide associated with Parkinson's disease risk, and certain fungicides, including pyraclostrobin, trifloxystrobin, famoxadone and fenamidone, produce transcriptional changes in vitro that are similar to those seen in brain samples from humans with autism, advanced age and neurodegeneration (Alzheimer's disease and Huntington's disease). These chemicals stimulate free radical production and disrupt microtubules in neurons, effects that can be reduced by pretreating with a microtubule stabilizer, an antioxidant, or with sulforaphane. Our study provides an approach to prospectively identify environmental chemicals that transcriptionally mimic autism and other brain disorders.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Anti-NeuN purified Antibody, from guinea pig, purified by affinity chromatography
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Supelco
Rotenone, PESTANAL®, analytical standard