MilliporeSigma
  • Protein kinase a-dependent phosphorylation of serine 119 in the proto-oncogenic serine/arginine-rich splicing factor 1 modulates its activity as a splicing enhancer protein.
Protein kinase a-dependent phosphorylation of serine 119 in the proto-oncogenic serine/arginine-rich splicing factor 1 modulates its activity as a splicing enhancer protein.
Genes & cancer (2012-03-07)
Anne Kristin Aksaas, Sissel Eikvar, Göran Akusjärvi, Bjørn S Skålhegg, Anne Katrine Kvissel
ABSTRACT
Serine/arginine-rich splicing factor 1 (SRSF1), previously designated SF2/ASF, belongs to a family of SR proteins that regulate constitutive and alternative splicing. SRSF1 expression is increased in tumors from several tissues and elicits changes in key target genes involved in tumor genesis. Several protein kinases phosphorylate SRSF1, which regulates its localization and function. It is previously reported that protein kinase A (PKA) phosphorylates SRSF1, but the importance of this modification is not well characterized. Here, we show that PKA phosphorylates SRSF1 on serine 119 in vitro. Phosphorylation of SRSF1 on this site enhanced the RNA binding capacity of SRSF1 in vivo and reduced the protein's capacity to activate splicing of the Minx transcript in vitro. We also confirm an interaction between SRSF1 and PKA Cα1 and demonstrate that this interaction is not dependent on serine 119 phosphorylation but requires active PKA Cα1. We conclude that PKA phosphorylation of SRSF1 at serine 119 regulates SFRS1-dependent RNA binding and processing but not its interaction with PKA.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Roche
Alkaline Phosphatase, solution, from bovine (calf) intestine, 2 units/μg protein, optimum pH 7.5-9.5
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder