MilliporeSigma
  • Deficiency of NPGPx, an oxidative stress sensor, leads to obesity in mice and human.
Deficiency of NPGPx, an oxidative stress sensor, leads to obesity in mice and human.
EMBO molecular medicine (2013-07-06)
Yi-Cheng Chang, Yu-Hsiang Yu, Jin-Yuh Shew, Wei-Jei Lee, Juey-Jen Hwang, Yen-Hui Chen, Yet-Ran Chen, Pei-Chi Wei, Lee-Ming Chuang, Wen-Hwa Lee
ABSTRACT
Elevated oxidative stress is closely associated with obesity. Emerging evidence shows that instead of being a consequence of obesity, oxidative stress may also contribute to fat formation. Nonselenocysteine-containing phospholipid hydroperoxide glutathione peroxidase (NPGPx) is a conserved oxidative stress sensor/transducer and deficiency of NPGPx causes accumulation of reactive oxygen species (ROS). In this communication, we show that NPGPx was highly expressed in preadipocytes of adipose tissue. Deficiency of NPGPx promoted preadipocytes to differentiate to adipocytes via ROS-dependent dimerization of protein kinase A regulatory subunits and activation of CCAAT/enhancer-binding protein beta (C/EBPβ). This enhanced adipogenesis was alleviated by antioxidant N-acetylcysteine (NAC). Consistently, NPGPx-deficient mice exhibited markedly increased fat mass and adipocyte hypertrophy, while treatment with NAC ablated these phenotypes. Furthermore, single nucleotide polymorphisms (SNPs) in human NPGPx gene, which correlated with lower NPGPx expression level in adipose tissue, were associated with higher body mass index (BMI) in several independent human populations. These results indicate that NPGPx protects against fat accumulation in mice and human via modulating ROS, and highlight the importance of targeting redox homeostasis in obesity management.
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, clone 10E9, clone 10E9, Upstate®, from mouse
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Roche
Cell Proliferation ELISA, BrdU (chemiluminescent)
Roche
First Strand cDNA Synthesis Kit for RT-PCR (AMV), sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included
Sigma-Aldrich
Sodium 4-Phenylbutyrate, A novel anti-neoplastic agent and transcriptional regulator.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra