MilliporeSigma
  • Neuropilin-1/GIPC1 signaling regulates alpha5beta1 integrin traffic and function in endothelial cells.
Neuropilin-1/GIPC1 signaling regulates alpha5beta1 integrin traffic and function in endothelial cells.
PLoS biology (2009-01-30)
Donatella Valdembri, Patrick T Caswell, Kurt I Anderson, Juliane P Schwarz, Ireen König, Elena Astanina, Francesca Caccavari, Jim C Norman, Martin J Humphries, Federico Bussolino, Guido Serini
ABSTRACT
Neuropilin 1 (Nrp1) is a coreceptor for vascular endothelial growth factor A165 (VEGF-A165, VEGF-A164 in mice) and semaphorin 3A (SEMA3A). Nevertheless, Nrp1 null embryos display vascular defects that differ from those of mice lacking either VEGF-A164 or Sema3A proteins. Furthermore, it has been recently reported that Nrp1 is required for endothelial cell (EC) response to both VEGF-A165 and VEGF-A121 isoforms, the latter being incapable of binding Nrp1 on the EC surface. Taken together, these data suggest that the vascular phenotype caused by the loss of Nrp1 could be due to a VEGF-A164/SEMA3A-independent function of Nrp1 in ECs, such as adhesion to the extracellular matrix. By using RNA interference and rescue with wild-type and mutant constructs, we show here that Nrp1 through its cytoplasmic SEA motif and independently of VEGF-A165 and SEMA3A specifically promotes alpha5beta1-integrin-mediated EC adhesion to fibronectin that is crucial for vascular development. We provide evidence that Nrp1, while not directly mediating cell spreading on fibronectin, interacts with alpha5beta1 at adhesion sites. Binding of the homomultimeric endocytic adaptor GAIP interacting protein C terminus, member 1 (GIPC1), to the SEA motif of Nrp1 selectively stimulates the internalization of active alpha5beta1 in Rab5-positive early endosomes. Accordingly, GIPC1, which also interacts with alpha5beta1, and the associated motor myosin VI (Myo6) support active alpha5beta1 endocytosis and EC adhesion to fibronectin. In conclusion, we propose that Nrp1, in addition to and independently of its role as coreceptor for VEGF-A165 and SEMA3A, stimulates through its cytoplasmic domain the spreading of ECs on fibronectin by increasing the Rab5/GIPC1/Myo6-dependent internalization of active alpha5beta1. Nrp1 modulation of alpha5beta1 integrin function can play a causal role in the generation of angiogenesis defects observed in Nrp1 null mice.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rab5 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin α3 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin α2 Antibody, serum, Chemicon®