MilliporeSigma
  • Bay 61-3606 Sensitizes TRAIL-Induced Apoptosis by Downregulating Mcl-1 in Breast Cancer Cells.
Bay 61-3606 Sensitizes TRAIL-Induced Apoptosis by Downregulating Mcl-1 in Breast Cancer Cells.
PloS one (2016-01-01)
So-Young Kim, Sang Eun Park, Sang-Mi Shim, Sojung Park, Kyung Kon Kim, Seong-Yun Jeong, Eun Kyung Choi, Jung Jin Hwang, Dong-Hoon Jin, Christopher Doosoon Chung, Inki Kim
ABSTRACT
Breast cancer cells generally develop resistance to TNF-Related Apoptosis-Inducing Ligand (TRAIL) and, therefore, assistance from sensitizers is required. In our study, we have demonstrated that Spleen tyrosine kinase (Syk) inhibitor Bay 61-3606 was identified as a TRAIL sensitizer. Amplification of TRAIL-induced apoptosis by Bay 61-3606 was accompanied by the strong activation of Bak, caspases, and DNA fragmentation. In mechanism of action, Bay 61-3606 sensitized cells to TRAIL via two mechanisms regulating myeloid cell leukemia sequence-1 (Mcl-1). First, Bay 61-3606 triggered ubiquitin-dependent degradation of Mcl-1 by regulating Mcl-1 phosphorylation. Second, Bay 61-3606 downregulates Mcl-1 expression at the transcription level. In this context, Bay 61-3606 acted as an inhibitor of Cyclin-Dependent Kinase (CDK) 9 rather than Syk. In summary, Bay 61-3606 downregulates Mcl-1 expression in breast cancer cells and sensitizes cancer cells to TRAIL-mediated apoptosis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Bak Antibody, NT, Upstate®, from rabbit
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK9