MilliporeSigma
  • Possibilities of retention prediction in fast gradient liquid chromatography. Part 3: Short silica monolithic columns.
Possibilities of retention prediction in fast gradient liquid chromatography. Part 3: Short silica monolithic columns.
Journal of chromatography. A (2015-08-05)
Pavel Jandera, Tomáš Hájek
ABSTRACT
We studied possibilities of prediction of the gradient elution data for alkylbenzenes, flavones and phenolic acids on two short octadecyl silica gel monolithic columns, namely a Chromolith Flash C18, 25×4.6mm, and a "new generation" Chromolith High Resolution C18, 50×4.6mm, in fast 1-2min gradients. With fixed short gradient times and varying gradient ranges of acetonitrile concentration in water, high flow rates of the mobile phase (3-5mL/min) could be used. The gradient elution data were predicted from four gradient models based on two-parameter and three-parameter isocratic retention equations. Various gradient retention models can be used for prediction of chromatograms and optimization of separation within a fixed gradient time. A two-parameter log-log model introduced in 1974 and a three-parameter model introduced in 1980 provided slightly more accurate prediction than the Linear Solvent Strength (LSS) semi-logarithmic two-parameter model, most frequently used in reversed-phase LC. A three-parameter model introduced in 1978 provided slightly improved accuracy of prediction of gradient data with respect to two-parameter models, in contrast to another, more recent three-parameter empirical model introduced in 2010 (which failed for gradients starting at a non-zero concentration of acetonitrile). Both a longer (5cm) and more efficient Chromolith HR column and a shorter (2.5cm) slightly less efficient Chromolith Flash column provide useful separations in fast gradients (1-2min) at high flow rates (3.5-5mL/min), especially in second dimension of two-dimensional LC×LC, in combination with HILIC separation on monolithic microcolumn in D1.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Pentylbenzene, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Rhein
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%