MilliporeSigma
  • Amplification of IL-21 signalling pathway through Bruton's tyrosine kinase in human B cell activation.
Amplification of IL-21 signalling pathway through Bruton's tyrosine kinase in human B cell activation.
Rheumatology (Oxford, England) (2015-03-01)
Sheau-Pey Wang, Shigeru Iwata, Shingo Nakayamada, Hiroaki Niiro, Siamak Jabbarzadeh-Tabrizi, Masahiro Kondo, Satoshi Kubo, Maiko Yoshikawa, Yoshiya Tanaka
ABSTRACT
B cells play an important role in the pathogenesis of autoimmune diseases. The role of Bruton's tyrosine kinase (Btk) in cytokine-induced human B cell differentiation and class-switch recombination remains incompletely defined. This study analysed the effect of Btk on human activated B cells. Purified B cells from healthy subjects were stimulated with B cell receptor (BCR) and other stimuli with or without a Btk inhibitor and gene expression was measured. The B cell line BJAB was used to assess Btk-associated signalling cascades. Phosphorylated Btk (p-Btk) in peripheral blood B cells obtained from 10 healthy subjects and 41 patients with RA was measured by flow cytometry and compared with patient backgrounds. IL-21 signalling, in concert with BCR, CD40 and BAFF signals, led to robust expression of differentiation- and class-switch DNA recombination-related genes and IgG production in human B cells, all of which were significantly suppressed by the Btk inhibitor. Although phosphorylation of STAT1 and STAT3 was induced by co-stimulation with IL-21, BCR and CD40, STAT1 phosphorylation in the nucleus, but not in the cytoplasm, was exclusively impaired by Btk blockade. High levels of p-Btk were noted in B cells of RA patients compared with controls and they correlated significantly with titres of RF among RF-positive patients. The findings elucidate a model in which Btk not only plays a fundamental role in the regulation of BCR signalling, but may also mediate crosstalk with cytokine signalling pathways through regulation of IL-21-induced phosphorylation of STAT1 in the nuclei of human B cells. Btk appears to have pathological relevance in RA.
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
SAFC
Glycine
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture