MilliporeSigma
  • Effects of diphenyl and p-chloro-diphenyl diselenides on feeding behavior of rats.
Effects of diphenyl and p-chloro-diphenyl diselenides on feeding behavior of rats.
Psychopharmacology (2015-01-08)
Cristiani F Bortolatto, Suélen O Heck, Bibiana M Gai, Vanessa A Zborowski, José S S Neto, Cristina W Nogueira
ABSTRACT
The searching for safe and effective antiobesity drugs has been the subject of intense research. Previous studies have shown several pharmacological applications of organoselenium compounds; however, their possible anorectic-like actions have not been investigated. This study aims to investigate the effects of (PhSe)2 and (p-ClPhSe)2 on feeding behavior of rats and their potential as weight-reducing agents. The effects of intraperitoneal administration of diselenides were investigated through the microstructural pattern of feeding behavior, behavioral satiety sequence (BSS), hypothalamic serotonin (5-HT) uptake, body weight, and epididymal fat content of male rats. Our findings demonstrated that food intake of fasted rats was reduced by both diselenides (1 and 10 mg/kg). Diphenyl diselenide [(PhSe)2] (1 mg/kg) and p-chloro-diphenyl diselenide [(p-ClPhSe)2] (10 mg/kg) decreased the frequency, mean duration, and mean size of meals compared with the control treatment. The BSS structure was preserved when organoselenium compounds (1 mg/kg) were administered, and it was associated to a displacement to the left when the resting period started indicating a satiating action. Inhibition of 5-HT uptake in the hypothalamus (∼20 %) was also found in rats treated with low doses of (PhSe)2 and (p-ClPhSe)2 (1 mg/kg). Treatments with a high dose of both diselenides (10 mg/kg) carried out for 7 days induced weight loss and epididymal fat reduction in sated rats. This study suggests that diselenides caused a satiating action in rats that could be partially explained by the inhibition of hypothalamic 5-HT uptake. These organoselenium compounds were potential weight-reducing agents when repeatedly administered.
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, ≥99%
Sigma-Aldrich
Lithium chloride, BioUltra, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Sibutramine hydrochloride monohydrate, ≥98% (HPLC), solid