MilliporeSigma
  • Agarose-assisted micro-contact printing for high-quality biomolecular micro-patterns.
Agarose-assisted micro-contact printing for high-quality biomolecular micro-patterns.
Macromolecular bioscience (2015-01-06)
Min Jee Jang, Yoonkey Nam
ABSTRACT
Micro-contact printing has been developed to print biomolecules, such as cell adhesive molecules, proteins, or DNAs, on a substrate, which can serve as experimental platforms for investigating biological issues and engineering biosensors. Despite the popularity of this method, it has been technically challenging to use a conventional stamp made of a hydrophobic polydimethoxysilane (PDMS) elastomer that often requires surface treatments to facilitate the inking and stamping of biomolecules. In this work, we proposed a new surface modification method for a PDMS stamp using agarose hydrogel and demonstrated the applications to the design of micro-patterned substrates with biomolecules. By using a simple bench-top dip-coating method with a commercial syringe pump to steadily pull out the stamp from boiled agarose solution, we coated an agarose layer on the stamp. It consequentially enhanced the transferability of ink molecules to the target substrate and the uniformity of printed patterns compared to the traditional methods for treating stamp surface such as surfactant coating and temporary oxidation with air plasma. In addition, this microstamping method was also used to produce patterns of proteins with the preservation of bioactivity, which could guide neuronal growth. Thus, we demonstrated the applicability to the interface designs of biochips and biosensors.
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
2-Propanol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Poly-L-lysine–FITC Labeled, mol wt 30,000-70,000
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Trichlorosilane, 99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O