MilliporeSigma
  • Anti-proliferative and pro-apoptotic effects of lichen-derived compound protolichesterinic acid are not mediated by its lipoxygenase-inhibitory activity.
Anti-proliferative and pro-apoptotic effects of lichen-derived compound protolichesterinic acid are not mediated by its lipoxygenase-inhibitory activity.
Prostaglandins, leukotrienes, and essential fatty acids (2015-05-13)
M Bessadóttir, F F Eiríksson, S Becker, M H Ögmundsdóttir, S Ómarsdóttir, M Thorsteinsdóttir, H M Ögmundsdóttir
ABSTRACT
Lipoxygenases (LOXs) and their products are involved in several biological functions and have been associated with carcinogenesis. Protolichesterinic acid (PA), a lichen metabolite, inhibits 5- and 12-LOX and has anti-proliferative effects on various cancer cell lines. Here, PA was shown to inhibit proliferation of multiple myeloma cells, RPMI 8226 and U266, and pancreatic cancer cells AsPC-1. Apoptosis was induced only in multiple myeloma cells. Cell-cycle associated changes in expression and sub-cellular localization of 5- and 12-LOX were not affected by PA but increased cytoplasmic localisation was found to accompany morphological changes at later stages. Assessment by mass spectrometry showed that PA entered the pancreatic cancer cells. However, effects on LOX metabolites were only evident after treatment with concentrations exceeding those having anti-proliferative effects and no effects were measurable in the myeloma cells. We conclude that the anti-proliferative and pro-apoptotic effects of PA are not mediated directly through inhibition of LOX activity.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Zileuton, ≥98% (HPLC)
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)