- Hydrolysis-Sensitive Dithiolethione Prodrug Micelles.
Hydrolysis-Sensitive Dithiolethione Prodrug Micelles.
Macromolecular bioscience (2015-06-24)
Urara Hasegawa, Naoya Tateishi, Hiroshi Uyama, André J van der Vlies
PMID26102371
ABSTRACT
Prodrug micelles carrying 5-(4-hydroxyphenyl)-3H-1,2-dithiole-3-thione (ADT-OH), a compound possessing chemopreventive properties, are prepared from amphiphilic block copolymers linking ADT-OH via an ester bond using glycine (PAM-PGlyADT) and isoleucine linkers (PAM-PIleADT). The release of ADT-OH from the PAM-PIleADT micelles is much slower than the PAM-PGlyADT micelles. The PAM-PGlyADT micelles show comparable toxicity with ADT-OH in different cancer cell lines, whereas the PAM-PIleADT micelles are not toxic up to 400 µM. This ADT-ester prodrug micelle approach enables to modulate the release rate of ADT-OH and thus might find application in cancer therapy and prevention.
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, neutral, free-flowing, Redi-Dri™