MilliporeSigma
  • Bovine serum albumin-sodium alkyl sulfates bioconjugates as drug delivery systems.
Bovine serum albumin-sodium alkyl sulfates bioconjugates as drug delivery systems.
Colloids and surfaces. B, Biointerfaces (2015-05-04)
M Benkő, N Varga, D Sebők, G Bohus, Á Juhász, I Dékány
ABSTRACT
Precipitation of bovine serum albumin (BSA) by anionic surfactants with alkyl chains of increasing lengths (octyl, decyl, dodecyl sulfates) was studied at room temperature, at pH 3.0, in isotonic sodium chloride solution. The particle size of albumin, the zeta potential, the surface charge and fluorescent properties of BSA-surfactant composites were investigated concerning addition of increasing amount of surfactant. The thermal stability of the systems was monitored by calorimetric analysis (DSC). The formation of the well-ordered structure in the self-assembly process in liquid phase was studied by XRD measurement. The structure of the precipitated BSA-surfactant nanocomposites was characterized by small-angle X-ray scattering (SAXS). Finally, ibuprofen (IBU) molecules were enclosed in BSA-surfactant bioconjugate systems and the release properties of the drug were investigated. It has been found out that, as a consequence to the increasing number of carbon atoms in the alkyl chains of the surfactant, the structure and the fluorescent properties of the aggregates formed can be controlled due to the increase in the hydrophobicity of BSA-surfactant composites. The bioconjugates are well applicable as carrier to realize controlled release of drug molecules.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium octyl sulfate, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O