MilliporeSigma
  • Soluble epoxide hydrolase is involved in the development of atherosclerosis and arterial neointima formation by regulating smooth muscle cell migration.
Soluble epoxide hydrolase is involved in the development of atherosclerosis and arterial neointima formation by regulating smooth muscle cell migration.
American journal of physiology. Heart and circulatory physiology (2015-10-11)
Qingjie Wang, Leijun Huo, Jinlong He, Wenshuang Ding, Hang Su, Dongping Tian, Carrie Welch, Bruce D Hammock, Ding Ai, Yi Zhu
ABSTRACT
Epoxyeicosatrienoic acids (EETs) have beneficial effects on cardiovascular disease. Soluble epoxide hydrolase (sEH) metabolizes EETs to less active diols, thus diminishing their biological activity. sEH inhibitors can suppress the progression of atherosclerotic lesions in animal models. However, the regulation of sEH in vascular smooth muscle cells (VSMCs) and role of sEH in patients with atherosclerosis have not been evaluated. We hypothesize that sEH in VSMCs plays a pivotal role in atherosclerosis and injury-induced neointima formation. In this study, sEH expression in human autopsy atherosclerotic plaque was determined by immunohistochemistry. In cultured rat and human VSMCs, the phenotypic switching marker and sEH expression induced by platelet-derived growth factor-BB (PDGF-BB) were examined by Western blot analysis. Carotid-artery balloon injury was performed after adenovirus-mediated overexpression of sEH or oral administration of a potent sEH inhibitor in Sprague-Dawley rats. sEH was highly expressed in VSMCs of the intima and media within human atherosclerotic plaque. In vitro, PDGF-BB upregulated the expression in VSMCs after transcription and promoted cell proliferation and migration; the latter effect could be largely attenuated by an sEH inhibitor. Adenovirus-mediated overexpression of sEH could mimic the effect of PDGF-BB and induce VSMC proliferation and migration. In vivo, the sEH inhibitor led to a significant decrease in injury-induced neointima formation in a rat carotid-artery injury model. These data establish the effect of sEH expression on atherosclerotic progression and vascular remodeling after injury, thus identifying a novel integrative role for sEH in VSMC phenotypic modulation and migration. Blocking sEH activity may be a potential therapeutic approach for ameliorating vascular occlusive disease.
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)