MilliporeSigma
  • Synthesis, characterization and observation of antisite defects in LiNiPO4 nanomaterials.
Synthesis, characterization and observation of antisite defects in LiNiPO4 nanomaterials.
Scientific reports (2015-06-20)
Murukanahally Kempaiah Devaraju, Quang Duc Truong, Hiroshi Hyodo, Yoshikazu Sasaki, Itaru Honma
ABSTRACT
Structural studies of high voltage cathode materials are necessary to understand their chemistry to improve the electrochemical performance for applications in lithium ion batteries. LiNiPO4 nanorods and nanoplates are synthesized via a one pot synthesis using supercritical fluid process at 450 °C for 10 min. The X-ray diffraction (XRD) analysis confirmed that LiNiPO4 phase is well crystallized, phase purity supported by energy dispersive spectroscopy (EDS) and elemental mapping by scanning electron transmission electron microscopy (STEM). For the first time, we have carried out direct visualization of atom-by-atom structural observation of LiNiPO4 nanomaterials using high-angle annular dark-field (HAADF) and annular bright-field (ABF) scanning transmission electron microscopy (STEM) analysis. The Rietveld refinement analysis was performed to find out the percentage of antisite defects presents in LiNiPO4 nanoplates and about 11% of antisite defects were found. Here, we provide the direct evidence for the presence of Ni atoms in Li sites and Li in Ni sites as an antisite defects are provided for understanding of electrochemical behavior of high voltage Li ion battery cathode materials.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Lithium acetylacetonate, 97%
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) chloride, 98%