MilliporeSigma
  • Advantages of the phosphatidylserine-recognizing peptide PSP1 for molecular imaging of tumor apoptosis compared with annexin V.
Advantages of the phosphatidylserine-recognizing peptide PSP1 for molecular imaging of tumor apoptosis compared with annexin V.
PloS one (2015-03-25)
Soyoun Kim, Sang Mun Bae, Junyoung Seo, Kiweon Cha, Meilan Piao, Sun-Ji Kim, Hye-Nam Son, Rang-Woon Park, Byung-Heon Lee, In-San Kim
ABSTRACT
A number of peptide-based indicators have been identified and reported as potential apoptosis probes, offering great promise for early assessment of therapeutic efficacy in several types of cancer. Direct comparison of the newly developed probes with previously used ones would be an important step in assessing possible applications. Here, we compared the newly identified peptide-based phosphatidylserine (PS) indicator PSP1 (CLSYYPSYC) with annexin V, a common probe for molecular imaging of apoptotic cells, with respect to PS binding kinetics, apoptotic cell-targeting ability, and the efficacy of homing to apoptotic tumor cells in a mouse model after treatment with the anticancer agent camptothecin. Our results indicate that PSP1 efficiently targeted apoptotic cells and generated apoptosis/tumor-specific signals after cancer treatment in the animal model, whereas a similar dose of annexin V showed weak signals. The formation of a stable complex of PSP1 with PS might be one reason for the efficient in vivo targeting. We suggest that PSP1 has potential advantages for in vivo apoptotic cell imaging and could serve as a platform for the development of de novo peptide-based probes for apoptosis.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Avanti
D-ribo-Phytosphingosine, Avanti Research - A Croda Brand 860499P, powder
Avanti
Liver PC, Avanti Research - A Croda Brand
Avanti
Liver PC, Avanti Research - A Croda Brand 840055P, powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%