MilliporeSigma
  • Systematic study of cell isolation from bovine nucleus pulposus: Improving cell yield and experiment reliability.
Systematic study of cell isolation from bovine nucleus pulposus: Improving cell yield and experiment reliability.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2015-06-04)
Juliana T Y Lee, Kenneth M C Cheung, Victor Y L Leung
ABSTRACT
Differences in matrix compositions in human nucleus pulposus (NP) clinical samples demand different cell isolation protocols for optimal results but there is no clear guide about this to date. Sub-optimal protocols may result in low cell yield, limited reliability of results or even failure of experiments. Cell yield, viability and attachment of cells isolated from bovine NP tissue with different protocols were estimated by cell counting, Trypan blue staining and cell culturing respectively. RNA was extracted from isolated cells and quantified by Nanodrop spectrometry and RT-qPCR. Higher collagenase concentration, longer digestion duration and pronase pre-treatment increased the cell yield. Cell viability remained high (<5% dead cells) even after 0.2% collagenase treatment for overnight. NP cells remained to have high ACAN, COL2A1, CDH2, KRT18, and KRT19 expression compared to muscle cells for different cell isolation conditions tested. Digestion by collagenase alone without the use of pronase could isolate cells from human degenerated NP tissue but clusters of cells were observed. We suggest the use of the disappearance of tissue as an indirect measure of cells released. This study provides a guide for researchers to decide the parameters involved in NP cell isolation for optimal outcome.
Product Description
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O