MilliporeSigma
  • Cyclodextrin Micellar LC for Direct Selective Analysis of Combined Dosage Drugs in Urine.
Cyclodextrin Micellar LC for Direct Selective Analysis of Combined Dosage Drugs in Urine.
Journal of chromatographic science (2014-12-30)
Hassan M Albishri, Deia Abd El-Hady, Roaa A Tayeb
ABSTRACT
Two cyclodextrin micellar liquid chromatographic methods were developed and applied to the simultaneous determination of bisoprolol/hydrochlorothiazide and atenolol/chlorthalidone combinations in urine matrices without sample pretreatment. These combined β-blockers and diuretics chemotherapies are commonly used in the treatment of hypertension and cardiovascular diseases. Hybrid isocratic mobile phases containing hydroxypropyl-β-cyclodextrin, sodium dodecyl sulfate, phosphate buffer and methanol on a Luna C18 column with 0.5 mL min(-1) flow rate and 25.0°C column temperature were used. The methods were sensitive enough for the determination of analytes at the therapeutic urine levels with limits of detections down to 1.0 µg mL(-1); relative standard deviations and recoveries were ranged between 1.5-4.4% and 98.00-109.52%, respectively. Urinary excretion studies showed that the detection of drugs is possible up to 24 h after their ingestion. The selective proposed separations with less consumption of organic solvents over the hitherto ones could be attributed to the four point competitive interactions among analysts, pseudostationary phases and a real stationary phase.
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis