MilliporeSigma
  • Levels of essential and non-essential metals in ginger (Zingiber officinale) cultivated in Ethiopia.
Levels of essential and non-essential metals in ginger (Zingiber officinale) cultivated in Ethiopia.
SpringerPlus (2015-03-20)
Yohannes Wagesho, Bhagwan Singh Chandravanshi
ABSTRACT
Ginger (Zingiber officinale Roscoe) is a common condiment for various foods and beverages and widely used worldwide as a spice. Its extracts are used extensively in the food, beverage, and confectionary industries in the production of products such as marmalade, pickles, chutney, ginger beer, ginger wine, liquors, biscuits, and other bakery products. In Ethiopia, it is among the important spices used in every kitchen to flavor stew, tea, bread and local alcoholic drinks. It is also chiefly used medicinally for indigestion, stomachache, malaria, fevers, common cold, and motion sickness. The literature survey revealed that there is no study conducted on the determination of metals in ginger cultivated in Ethiopia. Hence it is worthwhile to determine the levels of essential and non-essential metals in ginger cultivated in Ethiopia. The levels of essential (Ca, Mg, Fe, Zn, Cu, Co, Cr, Mn, and Ni) and non-essential (Cd and Pb) metals in ginger (Zingiber officinale Roscoe) cultivated in four different regions of Ethiopia and the soil where it was grown were determined by flame atomic absorption spectrometry. 0.5 g of oven dried ginger and soil samples were digested using 3 mL of HNO3 and 1 mL of HClO4 at 210°C for 3 h and a mixture of 6 mL aqua-regia and 1.5 mL H2O2 at 270°C for 3 h, respectively. The mean metal concentration (μg/g dry weight basis) ranged in the ginger and soil samples, respectively, were: Ca (2000-2540, 1770-3580), Mg (2700-4090, 1460-2440), Fe (41.8-89.0, 21700-46900), Zn (38.5-55.2, 255-412), Cu (1.1-4.8, 3.80-33.9), Co (2.0-7.6, 48.5-159), Cr (6.0-10.8, 110-163), Mn (184-401, 1760-6470), Ni (5.6-8.4, 14.1-79.3) and Cd (0.38-0.97, 0.24-1.1). The toxic metal Pb was not detected in both the ginger and soil samples. There was good correlation between some metals in ginger and soil samples while poor correlation between other metals (Fe, Ni, Cu). This study revealed that Ethiopian gingers are good source of essential metals and free from toxic metal Pb while containing negligible amount of Cd.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) nitrate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%