MilliporeSigma
Purifying Cytokinetic Cells from an Asynchronous Population.
Scientific reports (2015-08-12)
Einat Panet, Efrat Ozer, Tal Mashriki, Itay Lazar, Devora Itzkovich, Amit Tzur
ABSTRACT
Cytokinesis is an intensively studied process by which the cell cytoplasm divides to produce two daughter cells. Like any other aspect of cell cycle research, the study of cytokinesis relies heavily on cell synchronization. However, the synchronization of cells during cytokinesis is challenging due to the rapid nature of this process and the shortage of cell cycle blocking agents specifically targeting this phase. Here, we demonstrate the use of standard flow cytometry for directly isolating cytokinetic cells from an asynchronous population of normally proliferating cells. This approach is based on a cell cycle marker whose temporal proteolysis, in combination with DNA quantification or cell size approximation, distinguishes cells undergoing cytokinesis. Furthermore, by avoiding doublet discrimination, typically used in flow cytometry analyses, we were able to further increase selectivity, specifically purifying cells at late cytokinesis. Our method circumvents checkpoint activation, cell cycle arrest, and any other means of pre-synchronization. These qualities, as demonstrated for both unattached and adherent cells, enable high selectivity for cytokinetic cells despite their overall low abundance in an asynchronous population. The sorted cells can then be readily used for cell biological, biochemical, and genomic applications to facilitate cytokinesis and cell cycle research.
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-fd Bacteriophage−Biotin antibody produced in rabbit, ~3.5 mg/mL, IgG fraction of antiserum
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine