MilliporeSigma
  • Excision of Expanded GAA Repeats Alleviates the Molecular Phenotype of Friedreich's Ataxia.
Excision of Expanded GAA Repeats Alleviates the Molecular Phenotype of Friedreich's Ataxia.
Molecular therapy : the journal of the American Society of Gene Therapy (2015-03-12)
Yanjie Li, Urszula Polak, Angela D Bhalla, Natalia Rozwadowska, Jill Sergesketter Butler, David R Lynch, Sharon Y R Dent, Marek Napierala
ABSTRACT
Friedreich's ataxia (FRDA) is an autosomal recessive neurological disease caused by expansions of guanine-adenine-adenine (GAA) repeats in intron 1 of the frataxin (FXN) gene. The expansion results in significantly decreased frataxin expression. We report that human FRDA cells can be corrected by zinc finger nuclease-mediated excision of the expanded GAA repeats. Editing of a single expanded GAA allele created heterozygous, FRDA carrier-like cells and significantly increased frataxin expression. This correction persisted during reprogramming of zinc finger nuclease-edited fibroblasts to induced pluripotent stem cells and subsequent differentiation into neurons. The expression of FRDA biomarkers was normalized in corrected patient cells and disease-associated phenotypes, such as decreases in aconitase activity and intracellular ATP levels, were reversed in zinc finger nuclease corrected neuronal cells. Genetically and phenotypically corrected patient cells represent not only a preferred disease-relevant model system to study pathogenic mechanisms, but also a critical step towards development of cell replacement therapy.
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)
Sigma-Aldrich
Neurotrophin-3 human, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
NT-3 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
NT-3 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
BDNF human, Carrier free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), suitable for cell culture