MilliporeSigma
  • AIMP2/p38, the scaffold for the multi-tRNA synthetase complex, responds to genotoxic stresses via p53.
AIMP2/p38, the scaffold for the multi-tRNA synthetase complex, responds to genotoxic stresses via p53.
Proceedings of the National Academy of Sciences of the United States of America (2008-08-13)
Jung Min Han, Bum-Joon Park, Sang Gyu Park, Young Sun Oh, So Jung Choi, Sang Won Lee, Soon-Kyung Hwang, Seung-Hee Chang, Myung-Haing Cho, Sunghoon Kim
ABSTRACT
AIMP2/p38 is a scaffolding protein required for the assembly of the macromolecular tRNA synthetase complex. Here, we describe a previously unknown function for AIMP2 as a positive regulator of p53 in response to genotoxic stresses. Depletion of AIMP2 increased resistance to DNA damage-induced apoptosis, and introduction of AIMP2 into AIMP2-deficient cells restored the susceptibility to apoptosis. Upon DNA damage, AIMP2 was phosphorylated, dissociated from the multi-tRNA synthetase complex, and translocated into the nuclei of cells. AIMP2 directly interacts with p53, thereby preventing MDM2-mediated ubiquitination and degradation of p53. Mutations in AIMP2, affecting its interaction with p53, hampered its ability to activate p53. Nutlin-3 recovered the level of p53 and the susceptibility to UV-induced cell death in AIMP2-deficient cells. This work demonstrates that AIMP2, a component of the translational machinery, functions as proapoptotic factor via p53 in response to DNA damage.