MilliporeSigma
  • IL-17A induces hypo-contraction of intestinal smooth muscle via induction of iNOS in muscularis macrophages.
IL-17A induces hypo-contraction of intestinal smooth muscle via induction of iNOS in muscularis macrophages.
Journal of pharmacological sciences (2014-07-30)
Daisuke Mori, Nobumasa Watanabe, Osamu Kaminuma, Takahisa Murata, Takachika Hiroi, Hiroshi Ozaki, Masatoshi Hori
ABSTRACT
Intestinal inflammation causes disorder in bowel motility. Th17 cytokines are involved in intestinal inflammation. To understand the role of interleukin (IL)-17 in intestinal motility, we examined effects of IL-17A on contractile activities of organ-cultured ileum. Rat ileal smooth muscle strips were organ cultured with IL-17A. Muscle contraction was measured, and cells expressing inducible nitric oxide synthase (iNOS) were identified with immunohistochemistry. Creating Th17-transferred colitis model mice, in vivo effects of IL-17 on contractile activities, and iNOS mRNA expression in colonic smooth muscle were investigated. Treatment with IL-17A for 12 h and 3 days attenuated carbachol- and membrane depolarization-induced contractions in organ-cultured rat ileum. N(G)-Nitro-l-arginine methyl ester (100 μM), a nitric oxide synthase inhibitor, completely reversed the IL-17A-induced inhibition of contractile force. Ileal tissue cultured in the presence of IL-17A showed increased expression of iNOS mRNA and protein. Immunohistochemical analysis using an iNOS antibody revealed that iNOS protein was expressed on ED2-positive muscularis macrophages. The level of iNOS mRNA was also increased in inflamed colonic smooth muscle of Th17-transferred colitis model mice. In intestinal inflammation, IL-17A induces an intestinal motility disorder through iNOS expression in muscularis macrophages.
Product Description
Carbachol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free