MilliporeSigma
Decreased serum osmolality promotes ductus arteriosus constriction.
Cardiovascular research (2014-09-06)
Rika Aoki, Utako Yokoyama, Yasuhiro Ichikawa, Masataka Taguri, Shun Kumagaya, Ryo Ishiwata, Chiharu Yanai, Shujiro Fujita, Masanari Umemura, Takayuki Fujita, Satoshi Okumura, Motohiko Sato, Susumu Minamisawa, Toshihide Asou, Munetaka Masuda, Shiho Iwasaki, Shigeru Nishimaki, Kazuo Seki, Shumpei Yokota, Yoshihiro Ishikawa
ABSTRACT
At birth, dynamic changes occur in serum components and haemodynamics, such as closure of the ductus arteriosus (DA). A previous study demonstrated that, in full-term human neonates, serum osmolality decreased transiently after birth, but recovered over the next few days. However, the significance of this transient decrease in osmolality has never been addressed. The objective of the present study was to examine the role of changes in serum osmolality after birth in DA closure. We found that rats exhibited a similar transient hypoosmolality after birth. Hypotonic stimulation induced constriction of DA rings and increased Ca(2+) transient in DA smooth muscle cells, but not in the aorta. The hypoosmotic sensor transient receptor potential melastatin 3 (TRPM3) was highly expressed in the rat DA, and TRPM3 silencing abolished the Ca(2+) response to hypoosmolality. Pregnenolone sulfate stimulation of TRPM3 induced rat DA constriction ex vivo and in vivo. Furthermore, hypertonic fluid injection impaired rat DA closure. In humans, neonatal serum hypoosmolality was observed in relatively mature preterm infants (≥28 weeks). In extremely preterm infants (<28 weeks), however, this hypoosmolality was absent. Instead, a rapid increase in osmolality occurred thereafter. Such an increase was greater, in particular, among patent DA (PDA) patients. A transient decrease in serum osmolality may promote DA closure during the first few days of life. Adjusting serum osmolality to proper levels might help to prevent the onset of PDA, improving the therapeutic outcome in extremely preterm infants.
Sigma-Aldrich
KB-R7943, ≥98% (HPLC), powder
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Nicardipine hydrochloride, powder, ≥98%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material