MilliporeSigma
  • Cyclic AMP inhibits neuromuscular junction maturation mediated by intracellular Ca2+.
Cyclic AMP inhibits neuromuscular junction maturation mediated by intracellular Ca2+.
Neuroscience letters (2015-01-18)
Wei Song, Xiwan Albert Jin
ABSTRACT
The neuromuscular junction (NMJ) is established through initial contact of motor neuron axon with a skeletal muscle cell and the subsequent synaptic maturation. Previous studies have shown that cyclic AMP (cAMP) enhanced spinal neurons' survival and growth but inhibited synaptogenesis. Here, we find that elevating intracellular cAMP level of presynaptic neurons prevented NMJs from maturation both physiologically and morphologically. Importantly, cytosolic Ca(2+) is essential for the inhibitory effects of cAMP on NMJ maturation. We show that depletion of intracellular Ca(2+) store, rather than extracellular Ca(2+), abolished the cAMP-dependent inhibition of synaptic maturation. Taken together, we demonstrate that Ca(2+) released from intracellular Ca(2+) stores regulates neurotrophic actions on NMJ maturation.
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder